Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc27a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009512.2 | NP_033538.2 | 7 - 8 | 1793 | 88 | |
GenBank mRNA | AF072760.1 | - | 7 - 8 | 1582 | 88 | |
AI173562.1 | - | 3 - 4 | 88 | |||
AJ223959.1 | - | 7 - 8 | 1751 | 88 | ||
BC013272.1 | - | 7 - 8 | 1758 | 4193807 | 88 | |
BC013335.1 | - | 7 - 8 | 1758 | 4193567 | 88 | |
BC115421.2 | - | 7 - 8 | 1733 | 40093476 | 88 | |
BC145823.1 | - | 7 - 8 | 1733 | 40131126 | 88 | |
BC145825.1 | - | 7 - 8 | 1733 | 40131128 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |