Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr2c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011630.3 | NP_035760.1 | 12 - 13 | 1780 | 72 | |
GenBank mRNA | AK157305.1 | - | 12 - 13 | 1754 | 72 | |
AK220507.1 | - | 5 - 6 | 678 | 72 | ||
BC103683.1 | - | 10 - 11 | 1399 | 40047943 | 72 | |
BC103684.1 | - | 10 - 11 | 1399 | 40047942 | 72 | |
BC103686.1 | - | 10 - 11 | 1399 | 40047941 | 72 | |
S75970.1 | - | 12 - 13 | 1895 | 72 | ||
U11688.1 | - | 11 - 12 | 1442 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Nuclear Receptor Mammalian Gene Collection Mus musculus Probe spans exons |