Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ltbr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010736.3 | NP_034866.1 | 2 - 3 | 383 | 78 | |
GenBank mRNA | AI119599.1 | - | 2 - 3 | 383 | 78 | |
BC138590.1 | - | 2 - 3 | 246 | 8861610 | 78 | |
BC138591.1 | - | 2 - 3 | 246 | 8861611 | 78 | |
L38423.1 | - | 2 - 3 | 383 | 78 | ||
U29173.1 | - | 2 - 3 | 367 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |