Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172732.3 | NP_766320.2 | 3 - 4 | 409 | 107 | |
GenBank mRNA | AK041288.1 | - | 3 - 4 | 409 | 107 | |
EU339277.1 | - | 3 - 4 | 316 | 107 | ||
EU339280.1 | - | 3 - 4 | 316 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |