Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cidec | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301295.1 | NP_001288224.1 | 4 - 5 | 439 | 103 | |
NM_178373.3 | NP_848460.1 | 4 - 5 | 423 | 103 | ||
GenBank mRNA | AK080133.1 | - | 4 - 5 | 425 | 103 | |
BC099676.1 | - | 3 - 4 | 395 | 5043221 | 103 | |
M61737.1 | - | 4 - 5 | 439 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |