Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tet3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183138.2 | NP_898961.2 | 4 - 5 | 2437 | 65 | |
GenBank mRNA | HQ423151.1 | - | 4 - 5 | 2611 | 65 | |
HQ423152.1 | - | 3 - 4 | 2554 | 65 | ||
JX946278.1 | - | 5 - 6 | 3143 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |