Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Qrsl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081054.2 | NP_001074523.1 | 2 - 3 | 284 | 98 | |
GenBank mRNA | AK012351.1 | - | 2 - 3 | 285 | 98 | |
AK038801.1 | - | 2 - 3 | 293 | 98 | ||
AK041347.1 | - | 2 - 3 | 289 | 98 | ||
AK170225.1 | - | 2 - 3 | 229 | 98 | ||
BC070459.1 | - | 2 - 3 | 275 | 30605375 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |