Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smpdl3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133888.2 | NP_598649.1 | 2 - 3 | 397 | 81 | |
GenBank mRNA | AK136602.1 | - | 1 - 2 | 60 | 80 | |
AK154599.1 | - | 2 - 3 | 423 | 81 | ||
AK166253.1 | - | 2 - 3 | 373 | 81 | ||
BC009087.1 | - | 2 - 3 | 397 | 2810972 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |