Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ak2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033966.4 | NP_001029138.1 | 4 - 5 | 549 | 69 | |
NM_016895.4 | NP_058591.2 | 4 - 5 | 549 | 69 | ||
GenBank mRNA | AK010936.1 | - | 4 - 5 | 468 | 69 | |
AK010951.1 | - | 4 - 5 | 470 | 69 | ||
AK050133.1 | - | 4 - 5 | 510 | 69 | ||
AK077519.1 | - | 4 - 5 | 472 | 69 | ||
AK166976.1 | - | 4 - 5 | 460 | 69 | ||
AK167797.1 | - | 4 - 5 | 509 | 69 | ||
AK168056.1 | - | 4 - 5 | 472 | 69 | ||
AK168148.1 | - | 4 - 5 | 69 | |||
BC008610.1 | - | 4 - 5 | 459 | 3584786 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |