Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map7d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145970.1 | NP_001139442.1 | 6 - 7 | 1116 | 89 | |
GenBank mRNA | BC023774.1 | - | 6 - 7 | 1113 | 89 | |
CA750559.1 | - | 1 - 2 | 112 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |