Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grid2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008167.2 | NP_032193.1 | 5 - 6 | 783 | 70 | |
GenBank mRNA | AK141070.1 | - | 1635 | 70 | ||
BC139823.1 | - | 5 - 6 | 787 | 40141838 | 70 | |
D13266.1 | - | 5 - 6 | 783 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons |