Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172731.2 | NP_766319.2 | 7 - 8 | 2977 | 63 | |
GenBank mRNA | AK220153.1 | - | 8 - 9 | 3451 | 63 | |
BC042732.1 | - | 3 - 4 | 394 | 63 | ||
BC060664.1 | - | 7 - 8 | 2440 | 63 | ||
BC145502.1 | - | 8 - 9 | 2794 | 9054065 | 63 | |
BC150786.1 | - | 7 - 8 | 2926 | 9087697 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |