Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001160.3 | NP_001001160.1 | 5 - 6 | 1797 | 120 | |
NM_001289674.1 | NP_001276603.1 | 4 - 5 | 1628 | 120 | ||
NM_001289675.1 | NP_001276604.1 | 4 - 5 | 1622 | 120 | ||
GenBank mRNA | AK129466.1 | - | 3 - 4 | 1174 | 120 | |
AK147230.1 | - | 5 - 6 | 1689 | 120 | ||
AK147681.1 | - | 4 - 5 | 1487 | 120 | ||
BC061475.1 | - | 2 - 3 | 269 | 120 | ||
BC070445.1 | - | 4 - 5 | 1628 | 30536738 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |