Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emid1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080595.2 | NP_542162.1 | 6 - 7 | 575 | 118 | |
GenBank mRNA | AJ416093.1 | - | 6 - 7 | 575 | 118 | |
AK153702.1 | - | 7 - 8 | 693 | 118 | ||
BC130023.1 | - | 6 - 7 | 580 | 40140754 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |