Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007482.3 | NP_031508.1 | 6 - 7 | 774 | 81 | |
GenBank mRNA | AK149471.1 | - | 6 - 7 | 757 | 81 | |
AK171417.1 | - | 5 - 6 | 608 | 81 | ||
BC013341.1 | - | 6 - 7 | 751 | 4163626 | 81 | |
BC050005.1 | - | 6 - 7 | 788 | 3990246 | 81 | |
CT010173.1 | - | 6 - 7 | 672 | 81 | ||
U51805.1 | - | 6 - 7 | 813 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons |