Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhbdl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183163.2 | NP_898986.2 | 4 - 5 | 622 | 132 | |
GenBank mRNA | BC052650.1 | - | 4 - 5 | 1036 | 132 | |
BX511550.1 | - | 3 - 4 | 400 | 132 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |