Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0X00015420.mm.1 |
NM_001177973.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015420.mm.1 |
NM_001177973.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015420.mm.1 |
NM_001177974.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015420.mm.1 |
NM_001177974.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015420.mm.1 |
NM_001177975.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015420.mm.1 |
NM_001177975.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015420.mm.1 |
NM_001177976.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015420.mm.1 |
NM_001177976.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015420.mm.1 |
NM_008363.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015420.mm.1 |
NM_008363.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015420.mm.1 |
TC0X00002442.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00015424.mm.1 |
NM_001177973.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015424.mm.1 |
NM_001177973.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015424.mm.1 |
NM_001177974.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015424.mm.1 |
NM_001177974.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015424.mm.1 |
NM_001177975.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015424.mm.1 |
NM_001177975.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015424.mm.1 |
NM_001177976.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015424.mm.1 |
NM_001177976.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015424.mm.1 |
NM_008363.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015424.mm.1 |
NM_008363.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015424.mm.1 |
NR_030758.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015424.mm.1 |
NR_030758.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015424.mm.1 |
TC0X00002442.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00015418.mm.1 |
NM_001177973.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015418.mm.1 |
NM_001177973.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015418.mm.1 |
NM_001177974.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015418.mm.1 |
NM_001177974.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015418.mm.1 |
NM_001177975.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015418.mm.1 |
NM_001177975.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015418.mm.1 |
NM_001177976.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015418.mm.1 |
NM_001177976.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015418.mm.1 |
NM_008363.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015418.mm.1 |
NM_008363.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015418.mm.1 |
TC0X00002442.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00015413.mm.1 |
NM_001177973.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015413.mm.1 |
NM_001177973.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015413.mm.1 |
NM_001177974.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015413.mm.1 |
NM_001177974.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015413.mm.1 |
NM_001177975.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015413.mm.1 |
NM_001177975.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015413.mm.1 |
NM_001177976.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015413.mm.1 |
NM_001177976.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015413.mm.1 |
NM_008363.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015413.mm.1 |
NM_008363.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015413.mm.1 |
TC0X00002442.mm.1 |
MTA TC |