Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0X00004455.mm.1 |
NM_001142809.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00004455.mm.1 |
NM_001142809.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00004455.mm.1 |
NM_001142810.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00004455.mm.1 |
NM_001142810.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00004455.mm.1 |
NM_133987.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00004455.mm.1 |
NM_133987.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00004455.mm.1 |
TC0X00000690.mm.1 |
MTA TC |