Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnj14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145963.2 | NP_666075.1 | 1 - 2 | 129 | 107 | |
GenBank mRNA | AK030711.1 | - | 1 - 2 | 139 | 107 | |
AK044725.1 | - | 1 - 2 | 110 | 107 | ||
AK080732.1 | - | 1 - 2 | 118 | 107 | ||
BC029692.1 | - | 1 - 2 | 69 | 5363132 | 107 | |
BC031753.1 | - | 1 - 2 | 129 | 4503027 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |