Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053100.2 | NP_444330.2 | 2 - 3 | 1459 | 60 | |
GenBank mRNA | AK052763.1 | - | 2 - 3 | 194 | 60 | |
AK155303.1 | - | 2 - 3 | 1406 | 60 | ||
BC029283.1 | - | 2 - 3 | 1393 | 60 | ||
BC037065.1 | - | 2 - 3 | 744 | 60 | ||
BC060094.1 | - | 2 - 3 | 689 | 60 | ||
BC082298.1 | - | 2 - 3 | 1334 | 60 | ||
BC152343.1 | - | 2 - 3 | 795 | 40093266 | 60 | |
BC152344.1 | - | 2 - 3 | 795 | 40093269 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |