Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfxn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053196.3 | NP_444426.3 | 4 - 5 | 546 | 77 | |
GenBank mRNA | AF325261.1 | - | 4 - 5 | 504 | 77 | |
AK034875.1 | - | 546 | 77 | |||
AK149277.1 | - | 515 | 77 | |||
AK150722.1 | - | 4 - 5 | 497 | 77 | ||
BC019808.1 | - | 4 - 5 | 544 | 5043343 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |