Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172514.3 | NP_766102.1 | 8 - 9 | 1149 | 71 | |
GenBank mRNA | AK088711.1 | - | 8 - 9 | 1150 | 71 | |
AK170268.1 | - | 8 - 9 | 1146 | 71 | ||
BC117823.1 | - | 8 - 9 | 1082 | 40092863 | 71 | |
BC117824.1 | - | 8 - 9 | 1082 | 40092864 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |