Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps37d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199677.1 | NP_001186606.1 | 2 - 3 | 377 | 72 | |
GenBank mRNA | AY081953.2 | - | 2 - 3 | 377 | 72 | |
BC044789.1 | - | 2 - 3 | 280 | 3484221 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |