Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfswap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172276.3 | NP_758480.2 | 1 - 2 | 366 | 95 | |
GenBank mRNA | AK031872.1 | - | 1 - 2 | 357 | 95 | |
AK088182.1 | - | 1 - 2 | 357 | 95 | ||
AK140369.1 | - | 1 - 2 | 359 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |