Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnft2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109902.1 | NP_001103372.1 | 9 - 10 | 1311 | 116 | |
NM_172998.3 | NP_766586.2 | 10 - 11 | 1346 | 116 | ||
GenBank mRNA | AK046850.1 | - | 10 - 11 | 1347 | 116 | |
AK090256.1 | - | 9 - 10 | 1407 | 116 | ||
AK148930.1 | - | 9 - 10 | 1288 | 116 | ||
BC057302.1 | - | 9 - 10 | 1273 | 6827287 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |