Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnft2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109902.1 | NP_001103372.1 | 5 - 6 | 844 | 89 | |
NM_172998.3 | NP_766586.2 | 6 - 7 | 879 | 89 | ||
GenBank mRNA | AK046850.1 | - | 6 - 7 | 880 | 89 | |
AK090256.1 | - | 5 - 6 | 940 | 89 | ||
AK148930.1 | - | 5 - 6 | 821 | 89 | ||
BC057302.1 | - | 5 - 6 | 806 | 6827287 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |