Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnnm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031396.2 | NP_113573.2 | 2 - 3 | 1725 | 61 | |
GenBank mRNA | AF202994.1 | - | 2 - 3 | 951 | 61 | |
EF041508.1 | - | 2 - 3 | 621 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |