Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mybphl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026831.1 | NP_081107.1 | 6 - 7 | 958 | 84 | |
GenBank mRNA | AK004148.1 | - | 6 - 7 | 959 | 84 | |
BC089625.1 | - | 6 - 7 | 946 | 6756382 | 84 | |
BC132636.1 | - | 6 - 7 | 947 | 40130913 | 84 | |
BC132638.1 | - | 6 - 7 | 947 | 40130915 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |