Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028762.1 | NP_083038.1 | 6 - 7 | 944 | 83 | |
GenBank mRNA | AK053511.1 | - | 6 - 7 | 948 | 83 | |
AK146493.1 | - | 6 - 7 | 974 | 83 | ||
BC025619.1 | - | 6 - 7 | 944 | 5320947 | 83 | |
BC034010.1 | - | 6 - 7 | 910 | 4225239 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |