Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tatdn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175151.4 | NP_780360.1 | 5 - 6 | 376 | 220 | |
GenBank mRNA | AK010232.1 | - | 4 - 5 | 461 | 220 | |
AK042248.1 | - | 5 - 6 | 377 | 220 | ||
AK161972.1 | - | 5 - 6 | 461 | 220 | ||
BC034895.1 | - | 3 - 4 | 249 | 134 | ||
BC061187.1 | - | 5 - 6 | 373 | 30277245 | 220 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |