Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029270.2 | NP_083546.2 | 3 - 4 | 754 | 71 | |
NM_146161.3 | NP_666273.1 | 2 - 3 | 560 | 71 | ||
GenBank mRNA | AK080986.1 | - | 3 - 4 | 71 | ||
AK134817.1 | - | 1 - 2 | 378 | 71 | ||
BC025502.1 | - | 2 - 3 | 409 | 5321985 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |