Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172412.2 | NP_766000.1 | 2 - 3 | 452 | 68 | |
GenBank mRNA | AK010515.1 | - | 2 - 3 | 482 | 68 | |
AK049639.1 | - | 2 - 3 | 453 | 68 | ||
AK160959.1 | - | 2 - 3 | 498 | 68 | ||
BC056941.1 | - | 1 - 2 | 1151 | 68 | ||
BC083180.1 | - | 2 - 3 | 459 | 5707617 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |