Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtpbp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145147.5 | NP_660129.2 | 2 - 3 | 482 | 69 | |
GenBank mRNA | AJ293619.1 | - | 1 - 2 | 112 | 69 | |
AK144812.1 | - | 2 - 3 | 483 | 69 | ||
AK152225.1 | - | 2 - 3 | 483 | 69 | ||
AK152972.1 | - | 2 - 3 | 483 | 69 | ||
AK159462.1 | - | 2 - 3 | 455 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |