Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007651.3 | NP_031677.1 | 2 - 3 | 267 | 91 | |
GenBank mRNA | AA177409.1 | - | 2 - 3 | 155 | 91 | |
AK156510.1 | - | 2 - 3 | 174 | 91 | ||
AK160326.1 | - | 2 - 3 | 165 | 91 | ||
BC021310.1 | - | 2 - 3 | 181 | 5010267 | 91 | |
BC052905.1 | - | 2 - 3 | 132 | 30078163 | 91 | |
X97227.1 | - | 1 - 2 | 68 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |