Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oplah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153122.2 | NP_694762.1 | 17 - 18 | 2474 | 94 | |
GenBank mRNA | AK158391.1 | - | 17 - 18 | 2516 | 94 | |
AK164524.1 | - | 1 - 2 | 98 | 94 | ||
BC025120.1 | - | 4 - 5 | 572 | 4020691 | 94 | |
BC034522.1 | - | 17 - 18 | 2474 | 4236444 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |