Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bop1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013481.1 | NP_038509.1 | 5 - 6 | 670 | 65 | |
GenBank mRNA | AK079627.1 | - | 5 - 6 | 650 | 65 | |
AK129058.1 | - | 5 - 6 | 643 | 65 | ||
AK144095.1 | - | 5 - 6 | 681 | 65 | ||
AK167110.1 | - | 5 - 6 | 653 | 65 | ||
AK167225.1 | - | 5 - 6 | 654 | 65 | ||
AK167829.1 | - | 5 - 6 | 648 | 65 | ||
AK167878.1 | - | 5 - 6 | 658 | 65 | ||
AK168127.1 | - | 5 - 6 | 651 | 65 | ||
BC012693.1 | - | 5 - 6 | 644 | 4193123 | 65 | |
U77415.1 | - | 5 - 6 | 670 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |