Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Recql4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058214.3 | NP_478121.2 | 5 - 6 | 1193 | 137 | |
GenBank mRNA | AB039882.1 | - | 5 - 6 | 1066 | 137 | |
AB175741.1 | - | 5 - 6 | 1110 | 137 | ||
AK090260.1 | - | 6 - 7 | 794 | 137 | ||
BC064680.1 | - | 5 - 6 | 1167 | 137 | ||
BC131656.1 | - | 5 - 6 | 1106 | 137 | ||
BC131657.1 | - | 4 - 5 | 1293 | 137 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |