Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfrsf11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008764.3 | NP_032790.3 | 3 - 4 | 830 | 75 | |
GenBank mRNA | AB013898.1 | - | 3 - 4 | 830 | 75 | |
AK140244.1 | - | 3 - 4 | 653 | 75 | ||
AK146105.1 | - | 3 - 4 | 773 | 75 | ||
BC049782.1 | - | 3 - 4 | 647 | 6506550 | 75 | |
U94331.1 | - | 3 - 4 | 683 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |