Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163565.1 | NP_001157037.1 | 4 - 5 | 697 | 81 | |
NM_013643.2 | NP_038671.2 | 4 - 5 | 805 | 81 | ||
GenBank mRNA | AK038416.1 | - | 4 - 5 | 474 | 81 | |
BC051975.1 | - | 4 - 5 | 446 | 81 | ||
BC057339.1 | - | 3 - 4 | 967 | 81 | ||
BC079592.1 | - | 4 - 5 | 777 | 6817360 | 81 | |
S80329.1 | - | 1 - 2 | 641 | 81 | ||
U28216.1 | - | 4 - 5 | 558 | 81 | ||
U28217.1 | - | 4 - 5 | 816 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Phosphatase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |