Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008637.1 | NP_032663.1 | 2 - 3 | 85 | 60 | |
GenBank mRNA | AK011695.1 | - | 2 - 3 | 74 | 60 | |
AK088309.1 | - | 2 - 3 | 77 | 60 | ||
AK168312.1 | - | 2 - 3 | 88 | 60 | ||
BC098239.1 | - | 1 - 2 | 55 | 3469785 | 60 | |
D49956.1 | - | 2 - 3 | 85 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |