Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnah10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019536.1 | NP_062409.1 | 54 - 55 | 9361 | 76 | |
GenBank mRNA | AK147443.1 | - | 7 - 8 | 1992 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |