Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcolce | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008788.2 | NP_032814.2 | 2 - 3 | 281 | 66 | |
GenBank mRNA | AB008548.1 | - | 2 - 3 | 205 | 66 | |
AK142615.1 | - | 2 - 3 | 210 | 66 | ||
AK144392.1 | - | 2 - 3 | 282 | 66 | ||
AK146829.1 | - | 2 - 3 | 282 | 66 | ||
AK159430.1 | - | 2 - 3 | 269 | 66 | ||
AK159940.1 | - | 2 - 3 | 274 | 66 | ||
AK162524.1 | - | 2 - 3 | 285 | 66 | ||
X57337.1 | - | 2 - 3 | 248 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |