Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp3a13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007819.4 | NP_031845.1 | 3 - 4 | 320 | 75 | |
GenBank mRNA | AK136494.1 | - | 3 - 4 | 319 | 75 | |
AK136529.1 | - | 3 - 4 | 321 | 75 | ||
AK149572.1 | - | 3 - 4 | 321 | 75 | ||
BC046592.1 | - | 3 - 4 | 299 | 6308234 | 75 | |
X63023.1 | - | 3 - 4 | 296 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |