Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172462.4 | NP_766050.1 | 2 - 3 | 127 | 91 | |
GenBank mRNA | AK032872.1 | - | 2 - 3 | 141 | 91 | |
AK085077.1 | - | 1 - 2 | 113 | 91 | ||
BB623389.1 | - | 2 - 3 | 141 | 91 | ||
BC058553.1 | - | 2 - 3 | 125 | 91 | ||
BC094590.1 | - | 1 - 2 | 112 | 30616053 | 91 | |
BC139312.1 | - | 1 - 2 | 107 | 8862334 | 91 | |
BC139315.1 | - | 1 - 2 | 107 | 8862337 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |