Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133711.3 | NP_598472.2 | 1 - 2 | 232 | 70 | |
GenBank mRNA | AF395083.2 | - | 1 - 2 | 232 | 70 | |
AF530515.1 | - | 1 - 2 | 232 | 70 | ||
AK005612.1 | - | 1 - 2 | 232 | 70 | ||
BC048650.1 | - | 1 - 2 | 232 | 6772684 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |