Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wapal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004436.4 | NP_001004436.2 | 4 - 5 | 2126 | 141 | |
NM_001301330.1 | NP_001288259.1 | 4 - 5 | 2108 | 141 | ||
GenBank mRNA | AB167349.1 | - | 4 - 5 | 1932 | 141 | |
AK028167.1 | - | 2 - 3 | 230 | 141 | ||
AK129105.1 | - | 4 - 5 | 2126 | 141 | ||
BC139434.1 | - | 4 - 5 | 1881 | 8862456 | 141 | |
BC145625.1 | - | 4 - 5 | 1863 | 9054193 | 141 | |
CF161743.1 | - | 2 - 3 | 233 | 141 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |