Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vldlr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161420.1 | NP_001154892.1 | 7 - 8 | 1663 | 89 | |
NM_013703.2 | NP_038731.2 | 7 - 8 | 1663 | 89 | ||
GenBank mRNA | AK136838.1 | - | 7 - 8 | 1483 | 89 | |
BC013622.1 | - | 7 - 8 | 1099 | 3968213 | 89 | |
L33417.1 | - | 7 - 8 | 1087 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |