Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400002286.mm.1 |
NM_001114879.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400002286.mm.1 |
NM_001114879.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400002286.mm.1 |
NM_028945.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400002286.mm.1 |
NM_028945.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400002286.mm.1 |
TC1400000277.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400002284.mm.1 |
NM_001114879.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400002284.mm.1 |
NM_001114879.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400002284.mm.1 |
NM_028945.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400002284.mm.1 |
NM_028945.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400002284.mm.1 |
TC1400000277.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400002285.mm.1 |
NM_001114879.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400002285.mm.1 |
NM_001114879.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400002285.mm.1 |
NM_028945.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400002285.mm.1 |
NM_028945.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400002285.mm.1 |
TC1400000277.mm.1 |
MTA TC |