Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ept1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027652.2 | NP_081928.2 | 5 - 6 | 714 | 92 | |
GenBank mRNA | AK014530.1 | - | 4 - 5 | 585 | 92 | |
AK040745.1 | - | 5 - 6 | 624 | 93 | ||
AK048011.1 | - | 5 - 6 | 663 | 92 | ||
AK079325.1 | - | 5 - 6 | 690 | 92 | ||
AK122544.1 | - | 5 - 6 | 630 | 92 | ||
AK140416.1 | - | 4 - 5 | 577 | 92 | ||
BC106097.1 | - | 5 - 6 | 683 | 6485810 | 92 | |
BC115783.1 | - | 5 - 6 | 653 | 40104916 | 92 | |
BC117551.1 | - | 5 - 6 | 648 | 40104922 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |