Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0X00015843.mm.1 |
NM_001039373.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015843.mm.1 |
NM_010839.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015843.mm.1 |
TC0X00002476.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00015838.mm.1 |
NM_001039373.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015838.mm.1 |
TC0X00002476.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00015841.mm.1 |
NM_001039373.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015841.mm.1 |
NM_010839.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015841.mm.1 |
TC0X00002476.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00015840.mm.1 |
NM_001039373.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015840.mm.1 |
NM_010839.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015840.mm.1 |
TC0X00002476.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00015842.mm.1 |
NM_001039373.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015842.mm.1 |
NM_010839.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015842.mm.1 |
TC0X00002476.mm.1 |
MTA TC |